Learn More
Medchemexpress LLC HY-101382 5mg Medchemexpress, GR 103691 CAS:162408-66-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1013825MG
Encompass_Preferred
Medchemexpress, HY-101382 5mg GR 103691 CAS:162408-66-4 GR 103691 is a potent, selective dopamine D3 receptor antagonist with a Ki value of 0.4 nM. GR 103691 shows more than 100-fold selectivity for human dopamine human (h)D3 over hD4 and hD1 sites. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7842