Learn More
Abcam GR 127935 hydrochloride, 5-HT1B/1D receptor antagonist, 50MG
Supplier:  Abcam AB12052350MG
Encompass_Preferred
MW 534.06 g/mol, Purity >97%. Potent and selective 5-HT₁B/₁D receptor antagonist (pKi = 8.5 for both 5-HT₁D in rat and 5-HT₁D in guinea pig). 100-fold selectivity over 5-HT₁A, 5-HT₁E, and 5-HT₂C. Displays centrally mediated effects upon systemic administration.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7524