Learn More
Medchemexpress LLC Gr-46611 | 185259-85-2 | 99.9% | 377.48 g/mol | C23H27N3O2 | 1 MG

Supplier: Medchemexpress LLC HY1012481MG
GR-46611 is a small-molecule 5-HT1D receptor agonist used in pharmacological research, including studies of bladder hyperactivity and certain leukemia models. It is supplied as a high-purity research compound in multiple milligram pack sizes for preclinical and in vitro applications.
- Acts as a 5-HT1D receptor agonist suitable for receptor pharmacology studies.
- High purity (99.86%).
- Molecular weight 377.48 g/mol.
- Chemical formula C23H27N3O2.
- Available in small milligram quantities for dosing and assay work.
- Storage: room temperature for shipping; 4°C for long-term storage; in solvent, -80°C for 6 months or -20°C for 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.