Learn More
Abcam GR 55562 dihydrochloride, 5-HT1B / 5-HT1D receptor antagonist, 50MG
Supplier:  Abcam AB12051350MG
Encompass_Preferred
MW 448.4 Da, Purity >99%. Selective, competitive 5-HT1B/ 5-HT1D receptor antagonist (pKi values are 7.4 and 6.2, respectively). Inhibits 5-HT evoked contraction in pulmonary resistance arteries (pEC10 = 7.26).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7534