Learn More
Medchemexpress LLC GR 55562 hydrochloride | 172854-55-6 | 98.0% | 411.92 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW01086910MG
Encompass_Preferred
GR 55562 hydrochloride is a selective 5-HT1B receptor antagonist. It can be used for the research of nerve disease. This product is for research use only and not sold to patients.
  • Selective 5-HT1B receptor antagonist.
  • Applicable for research on nerve disease.
Catalog No. 50-004-53058