Learn More
Genetex Inc Glucocorticoid Receptor Monoclonal Antibody [HMV304] HistoMAX™ (GTX639927) GeneTex - IHC-Optimized Antibody
SDP
Supplier:  Genetex Inc GTX639927
Encompass_Preferred
The Glucocorticoid Receptor antibody [HMV304] HistoMAX™ (GTX639927) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human adrenal gland tissue using GTX639927 Glucocorticoid Receptor antibody [HMV304] HistoMAX. Moderate to strong GR positivity of medullary cells while adrenocortical cells are largely GR negative.
Catalog No. NC3669294