Learn More
Medchemexpress LLC GR24 (Strigolactone GR24) | 76974-79-3 | 99.9% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY129038
Encompass_Preferred
(Rac)-GR24 (Strigolactone GR24) is a synthetic analog of strigolactone, a plant hormone. It mimics the natural germination stimulus of parasitic plant seeds, promoting seed germination even without host plants, thus helping reduce parasitic seeds in the soil. It is utilized in agricultural research.
  • Promotes primary root elongation in Arabidopsis.
  • Inhibits hypocotyl elongation and lateral root density.
Catalog No. 50-003-41817