Learn More
Sigma Aldrich Fine Chemicals Biosciences Grace Bio-Labs ProPlate MP1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GBL2063841EA
Encompass
Patented ProPlate(R) technology for high throughput robotic processing of microarrays.The superstructures come in a permanently affixed well format--the ProPlate MP(TM) or a removable well format--the flexPlate MP(TM). The ProPlate MP(TM) SBS compliant microtiter plate features a SecureSeal(TM) Plus adhesive bonding system applied to a bottomless 96-round well black polystyrene microtiter plate. Guaranteed flat SBS compliant mircrotiter plate format which enables robotic processing of assays Water-tight seal between superstructure and substrate Compatible with glass or plastic substrates Available in 96 round or square or 384 square well formats
Catalog No. 50-290-1584