Learn More
Medchemexpress LLC Gracillin | 19083-00-2 | 885.04 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN07065MG
Encompass_Preferred
Gracillin is a steroidal saponin extracted from plant roots with anti-tumor properties. It can induce apoptosis and autophagy in cancer cells.
  • Steroidal saponin
  • Anti-tumor properties
  • Induces cancer cell apoptosis and autophagy
  • Shows broad-spectrum inhibitory effects on cancer cell viability
  • Disrupts mitochondrial function, inhibits ATP production, activates AMPK, and inhibits oxygen consumption rate
  • Induces ROS generation
  • Blocks cell cycle in G1 phase and induces apoptosis and autophagy
  • Inhibits tumor growth in xenograft models and patient-derived xenograft tumors
Catalog No. 50-004-15465