Learn More
Chemglass Life Sciences Cylinder, Economy, 50mL, Graduated, Double Scale
SDP
Supplier:  Chemglass Life Sciences CG824650
Encompass_Preferred
Calibrated "To Contain". A less costly cylinder, designed for those institutions performing many general laboratory procedures. No bumper guards are supplied.
Catalog No. NC0175351