Learn More
Roboz Surgical Graefe Tissue Forceps
SDP
Supplier:  Roboz Surgical RS5159
Encompass
Graefe tissue forceps with 1X2 teeth. The tips are fully curved. These forceps are lighter and finer than the regular Graefe forceps. Their fine tips are 0.5mm wide. The overall length of this instrument is 4".
Catalog No. NC0735136
 Restricted Item