Learn More
AdipoGen Gram’s Decolorizer Solution (250 ml)
SDP
Supplier:  AdipoGen CDXG0062L250
Encompass_Preferred
Gram’s Decolorizer Solution (250 ml), Adipogen Life Sciences. Chemical. CAS: 37348-17-7 (mixture). Formula: . MW: . Synthetic. Gram Decolorizer Solution is a reagent recommended for use in qualitative procedures to differentiate gram-negative from gram-positive organisms. Mixture used to decolorize Gram-negative bacteria. The Gram staining allows a fast differentiation of bacteria in Gram-positive and Gram-negative. Bacteria will be stained with Gram's crystal violet solution - an aniline dye - in the first step. After the treatment with iod solution, a dye-iod complex will form. During the decolorizing step, this complex stays in the multilayer mureine structures of the Gram-positive bacteria and they will appear blue. Gram-negative bacteria have a monolayer mureine structure only, the dye-iod complex does not stay bound to the cellwall, they will be decolorized. Gram-negative bacteria will be counterstained by safranin solution and appear orange.
Catalog No. 50-168-7616