Learn More
Aviva Systems Biology Corp GRAMD2B Antibody - middle region (ARP49782_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49782_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GRAMD3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human GRAMD3. Alternate names: FLJ21313, NS3TP2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-449