Learn More
Selleck Chemical LLC Gramine 25mg 87-52-5
SDP
Supplier:  Selleck Chemical LLC S230425MG
Encompass_Preferred
Gramine (Donaxine) is a natural indole alkaloid isolated from giant reed, acts as an active adiponectin receptor (AdipoR) agonist, with IC50 of 3.2 ¿M and 4.2 ¿M for AdipoR2 and AdipoR1, respectively. Gramine is also a human and mouse ?2-Adrenergic receptor (?2-AR) agonist. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-823-9