Learn More
Apexbio Technology LLC Granisetron HCl 107007-99-8 100mg
SDP
Supplier:  Apexbio Technology LLC A1295100
Encompass_Preferred
Granisetron HCl (CAS 107007-99-8) is a selective antagonist of the serotonin 5-hydroxytryptamine type 3 (5-HT sub 3 /sub ) receptor By competitively inhibiting 5-HT sub 3 /sub receptor-mediated signaling Granisetron HCl suppresses serotonin-induced activation of the vomiting reflex pathway in both central and peripheral nervous systems This pharmacological activity underlies its utility in preventing and investigating the mechanisms of chemotherapy-induced and postoperative nausea and vomiting Granisetron HCl is widely employed in preclinical and clinical studies to elucidate 5-HT sub 3 /sub receptor function and to explore antiemetic strategies
Catalog No. 50-000-96938