Learn More
Selleck Chemical LLC Granisetron HCl 25mg 107007-99-8
SDP
Supplier:  Selleck Chemical LLC S134525MG
Encompass_Preferred
Granisetron is a serotonin 5-HT3 receptor antagonist,used to treat chemotherapy-induced nausea and vomiting. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-652-9