Learn More
Medchemexpress LLC Granisetron Hydrochloride | 107007-99-8 | 99.9% | C18H25ClN4O | 100 MG

Supplier: Medchemexpress LLC HYB0071A100MG
Granisetron Hydrochloride is a serotonin 5-HT3 receptor antagonist used as an antiemetic to treat nausea and vomiting following chemotherapy. This product is for research use only.
- Serotonin 5-HT3 receptor antagonist
- Used as an antiemetic for chemotherapy-induced nausea and vomiting
- White to off-white solid appearance
- Purity of 99.9%
- Molecular weight of 348.87
- CAS number 107007-99-8
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.