Learn More
Medchemexpress LLC Granisetron hydrochloride (BRL 43694A) | 107007-99-8 | MFCD01747034 | 99.9% | C18H25ClN4O | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB0071A
Encompass_Preferred
Granisetron hydrochloride is an antiemetic medication and a selective serotonin 5-HT3 receptor antagonist. It functions by blocking serotonin at 5-HT3 receptors, thereby suppressing nausea and vomiting, particularly those induced by chemotherapy and radiotherapy.
  • Selective serotonin 5-HT3 receptor antagonist
  • Used as an antiemetic to treat nausea and vomiting
  • Helps suppress chemotherapy- and radiotherapy-induced nausea and vomiting
  • FDA approved for various antiemetic applications
  • Can prevent and treat postoperative nausea and vomiting
Catalog No. 50-003-59039