Learn More
Medchemexpress LLC Granzyme A (recombinant human) | >95.0% | 10 UG

Supplier: Medchemexpress LLC HYP7637710UG
Recombinant human granzyme A (GZMA) expressed in HEK293 cells with a C-terminal His tag. Supplied as a lyophilized powder for research use; the proenzyme requires activation to exhibit serine protease activity and cleaves lysine/arginine-containing substrates involved in immune cell cytotoxicity.
- Species: Human
- Expression system: HEK293 cells
- Tag: C-terminal His
- Purity: >95.0% by reducing SDS-PAGE
- Format: Lyophilized powder, 10 μg pack
- Endotoxin: <1 EU/μg (LAL)
- Specific activity: >5,000 pmol/min/μg after activation
- Storage: Store at -20°C; aliquot for long-term storage
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.