Learn More
Biolegend Alexa Fluor 700 anti-human/mouse Granzyme B Recombinant
Supplier:  Biolegend 372222
Encompass_Preferred
Alexa Fluor 700 anti-human/mouse Granzyme B Recombinant [QA16A02]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: ICFC; Size: 100 tests
Catalog No. NC1830154