Learn More
Trajan Scientific GRAPHITE FERRULES 1/16 10/PK
Supplier:  Trajan Scientific 211165/DUP
Encompass_Preferred
0.5mm ID Graphite Ferrule for Agilent GC Inlet,10/pack,100% graphite ferrule,upper temperature limit 450 deg.C,Special (short) style for Agilent inlet systems. Soft; graphite is recommended for use with fused silica and glass
Catalog No. NC9772436