Learn More
Glass Expansion Graphite Sampler Cone Gasket for Thermo Scientific Element 1/Element 2/Neptune/Neoma ICP-MS (1042620)
SDP
Supplier:  Glass Expansion TF5001
Encompass_Preferred
Graphite Sampler Cone Gasket (1042620) for Thermo/Finnigan Neoma MC-ICP-MS. Use a new gasket each time the cone is installed, even if it is the same cone.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7717