Learn More
Perkin Elmer US LLC Graphite/Vespel Ferrule, 0.0625 in
Supplier:  Perkin Elmer US LLC 09920511
Encompass_Preferred
Graphite/Vespel ferrules are 15% graphite and 85% polyimide. They are the best choice for glass columns and polyimide-coated, fused-silica capillary columns and can usually be removed easily from columns. Ferrule Type: No Hole Ferrule Inner Diameter: 0.0625 in Fitting Size: 0.0625 in
Catalog No. 50-211-2710