Learn More
Perkin Elmer US LLC Graphite/Vespel Ferrules - 0.125 in Nut x 0.0625 in I.D., Pkg. 10
Supplier:  Perkin Elmer US LLC 09920301
Encompass_Preferred
Recommended for use with Detectors/Injectors at temperatures up to 350 °C.
Catalog No. 50-211-7125