Learn More
Medchemexpress LLC Graphitic carbon nitride | 143334-20-7 | MFCD00040542 | 97.0% | C3N4 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW5400001G
Encompass_Preferred
Graphitic carbon nitride is a polymeric, metal-free photocatalyst (empirical formula near C3N4) used in visible-light photocatalysis and semiconductor materials research. It is supplied as a powder for laboratory research and exhibits semiconductor absorption in the blue-visible region, making it suitable for photocatalytic and energy-conversion studies.
  • Metal-free photocatalyst suitable for visible-light applications.
  • Semiconductor absorption near 460 nm for photoactive processes.
  • High purity for consistent experimental performance.
  • Powder form for easy dispersion and material processing.
  • Applicable to photocatalysis, water treatment, and energy conversion research.
  • Intended for research use only.
Catalog No. 50-002-84346