Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Scale with Cross Line Microscope Graticle NE7, 23 mm, 10 mm with 0.1 mm spacing
SDP

Supplier:  Electron Microscopy Sciences 6801323

Encompass_Preferred

Used as a horizontal and a vertical scale, they are especially useful when interested in measurement in different axis. Horizontal micrometer scale 10mm long with crosslines at 0.1mm spacing. Surface chrome image.
Size: 23 mm
Divisions: 10 mm with 0.1 mm spacing
Surface: Chrome image

Catalog No. 50-192-7993


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.