Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Cross Lines Graticle NE8, 27 mm, Nominal line width 0.020 mm
SDP

Supplier:  Electron Microscopy Sciences 6798327

Encompass_Preferred

Used as NE50 but for measurements in two directions and for sighting and alignment. Image covers the entire field of view.
Size: 27 mm
Divisions: Nominal line width 0.020 mm
Surface: Chrome image

Catalog No. 50-192-7980


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.