Learn More
Medchemexpress LLC Graveoline | 485-61-0 | MFCD00236478 | 99.9% | 279.29 g/mol | C17H13NO3 | 5MG
SDP
Supplier:  Medchemexpress LLC HY125385MG
Encompass_Preferred
Graveoline (CAS 485-61-0) is a quinolinone alkaloid (molecular formula C17H13NO3, molecular weight 279.29 g/mol) provided as a high-purity solid for research and analytical use. It is studied for antifungal, antiparasitic, anti-inflammatory, and antitumor activities, and is intended for use as a reference standard or analytical reagent. Certificate of analysis reports purity by HPLC as 99.88%.
  • High purity suitable for analytical applications.
  • Documented purity with certificate of analysis.
  • Characterized by CAS registry number 485-61-0.
  • Provided as a stable solid for storage and handling.
  • Appropriate for use as a reference standard in HPLC and other analyses.
Catalog No. 50-002-37438