Learn More
Anton Paar Gravitational free-fall dispersion with automated feeding for optimal results. One-click access enables easy cleaning, while a built-in drawer allows sample recovery. Designed for use with the Litesizer DIA series.
Supplier:  Anton Paar 261788
Encompass_Preferred
FREE FALL DISPERSION UNITDispersion unit using gravitational fall.Suitable for free-flowing and non-agglomerating particles up to16000 µm (sample dependent).Measurement cell: 4 mm gap (default). Optional: 8 mm gap.Sample funnel volume: 600 mL (default). Optional: 150 mL.Superior protection against sample spread.Automated feeding rate.The sample is retrieved after the measurement.
Catalog No. 50-005-80536
Request a Quote
Please contact your sales representative for a quote or further ordering instructions.