Learn More
Medchemexpress LLC HY-15298A 10mg Medchemexpress, Grazoprevir potassium salt CAS:1206524-86-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15298A10MG
Encompass_Preferred
Medchemexpress, HY-15298A 10mg Grazoprevir potassium salt CAS:1206524-86-8 Grazoprevir potassium salt (MK-5172 potassium salt) is a selective inhibitor of Hepatitis C virus NS3/4a protease with broad activity across genotypes and resistant variants, with Kis of 0.01 nM (gt1b), 0.01 nM (gt1a), 0.08 nM (gt2a), 0.15 nM (gt2b), 0.90 nM (gt3a), respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9113