Learn More
Medchemexpress LLC Grazoprevir potassium salt | 1206524-86-8 | C38H49KN6O9S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY15298A5MG
Encompass_Preferred
Grazoprevir potassium salt (MK-5172 potassium salt) is a selective inhibitor of Hepatitis C virus NS3/4a protease, demonstrating broad activity across various genotypes and resistant variants. This compound also inhibits SARS-CoV-2 3CLpro activity and is intended for research use only.
  • Selective inhibitor of Hepatitis C virus NS3/4a protease
  • Broad activity across genotypes and resistant variants
  • Inhibits SARS-CoV-2 3CLpro activity
  • Features Ki values such as 0.01 nM (gt1b) and 0.01 nM (gt1a)
Catalog No. 50-003-72514