Learn More
Medchemexpress LLC Green DND-26 | 220524-71-0 | 99.77% | 362.23 | 50 MG
Supplier: Medchemexpress LLC HYD129650MG
Green DND-26 is a green fluorescently labeled lysosomal probe with a maximum excitation/emission wavelength of 504/511 nm. Its structure consists of a fluorescein group and linked weak bases, allowing it to freely cross the cell membrane and accumulate in spherical organelles. It is used for observing the internal biosynthesis and related pathogenesis of lysosomes.
- Green fluorescently labeled lysosomal probe.
- Maximum excitation/emission wavelength: 504/511 nm.
- Composed of a fluorescein group and linked weak bases.
- Can freely cross the cell membrane and gather on spherical organelles.
- Suitable for observing internal biosynthesis and related pathogenesis of lysosomes.