Learn More
Alkali Scientific Green Dye 20mL 10% Vial 10/Pk
Supplier: Alkali Scientific GDV000
Green Dye, 20mL Sterile Vial (10%), 10/Pk is a visualization solution designed for aseptic process simulations, operator training, and media fill validations in controlled cleanroom environments. The 10% dye concentration provides strong color contrast, allowing operators to clearly observe fluid flow, transfer accuracy, and procedural integrity during validation exercises. Packaged in sterile 20mL vials, the solution is suitable for pharmaceutical, biotechnology, and GMP regulated facilities where reliable process verification is essential. The dye supports realistic simulation of liquid handling workflows without introducing microbial contamination or chemical interference. This makes it a practical tool for validating aseptic technique, performing equipment qualification, and improving operator confidence in sterile fluid handling procedures.