Learn More
Montana Molecular Nucleus-Targeted mNeonGreen (Green H2BPaint) Expression Kit - Big Sky BacMam, 5mL - Montana Molecular
SDP
Supplier:  Montana Molecular P1020G005
Encompass_Preferred
Bright, green fluorescent nuclear label for live-cell imaging—no fixation, staining, or cell lysis required. Visualize nuclei and cellular dynamics in real time, over hours to days, in living cells. Delivered via Montana Molecular’s Big Sky BacMam vector for enhanced expression across a wide range of cell types, including primary cultures and iPSCs. Targeted to nucleus via NLS. Ideal for dynamic, non-destructive studies of cellular phenotypes, cell tracking, and high-content imaging.
Catalog No. NC3787526