Learn More
Montana Molecular Omegasome-targeted mNeon Green (Green OmegaPaint) Expression Kit - Big Sky BacMam, 5mL - Montana Molecular
SDP
Supplier:  Montana Molecular P1090G005
Encompass_Preferred
Bright, green fluorescent omegasomal label for live-cell imaging—no fixation, staining, or cell lysis required. Visualize omegasomal morphology and spatial dynamics in real time over hours to days in living cells. Delivered using Montana Molecular’s Big Sky BacMam vector for robust expression across a wide range of cell types, including primary cultures and iPSCs. Omegasomal targeting is achieved via the double FYVE-containing protein 1 (ZNFN2A1) domain. Ideal for dynamic, non-destructive studies of cellular phenotypes, omegasome organization, and high-content imaging.
Catalog No. NC3787565