Learn More
Leica Biosystems Leica Biosystems Green Snowcoat Microscope Slides - 1,440 slides per case
Supplier:  Leica Biosystems 3800330
Encompass
Green Snowcoat Microscope Slides are color-coded Snowcoat-surface slides intended for histology and general microscopy workflows. Supplied in bulk cases to support high-volume laboratory processing and easy slide identification.
  • Green color for quick visual identification.
  • 1,440 slides per case for high-volume supply.
  • Snowcoat surface to aid consistent specimen spreading and handling.
  • Suitable for routine histology staining and microscopy applications.
  • Bulk packaging reduces reordering frequency and handling time.
Catalog No. NC3237839