Learn More
Medchemexpress LLC Green Tea Extract | 5 G

Supplier: Medchemexpress LLC HYN132295G
Green Tea Extract is derived from green tea, containing 30%-98% Tea Polyphenol. It is intended for research use only.
- Solid appearance, brown to reddish brown in color.
- Recommended storage at room temperature, kept dry and cool.
- For solutions, store at -80°C for 1 year or -20°C for 6 months.
- Soluble in DMSO at 100 mg/mL.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.