Learn More
Medchemexpress LLC GRGDSP | 91037-75-1 | 99.95% | 587.58 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYP0290100MG
Encompass_Preferred
GRGDSP is a synthetic linear RGD peptide and an integrin inhibitor. It can inhibit the adherence of tumor cells to endothelial cells of blood vessels and limit metastasis.
  • Integrin inhibitor
  • Inhibits adherence of tumor cells to endothelial cells
  • Limits metastasis
  • Used in integrin research as a soluble RGD-based peptide
  • Can be used to modify cardiovascular implants to promote endothelialization
Catalog No. 50-004-24932