Learn More
Medchemexpress LLC GRGDSP TFA | 98.0% | 701.61 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP0290A10MG
Encompass_Preferred
GRGDSP TFA is an integrin inhibitor. This synthetic linear RGD peptide is used as a soluble integrin-blocking agent in research.
  • Inhibits the adherence of tumor cells to endothelial cells of blood vessels.
  • Limits tumor metastasis.
  • Utilized with other RGD peptides in integrin research.
  • Modifies cardiovascular implant surfaces to promote endothelialization.
Catalog No. 50-004-24933