Learn More
Carbosynth LLC. H-Gly-Arg-Gly-Asp-Ser-Pro-Cys-OH, 1mg, MW: 690.74, Molecular formula: C23H42N10O11S, CAS# 91575-26-7
SDP
Supplier:  Carbosynth LLC. RGD3506PI1MG
Encompass_Preferred
This item has a minimum order quantity of 4 per supplier requirements
Application: Thiolated Cell Adhesion Peptide, Storage Temperature: -20C, References: M.K. Nguyen, et al., J Mater Chem B, 5(3), 485 (2017)., C. Tao, et al., Colloids Surf B Biointerfaces,122, 439 (2014)., T.A.Petrie et al., J. Cell. Mol. Med., 13, 2602 (2009).
Catalog No. 50-168-6926