Learn More
Hampton Research HR2-930-A3/Grid Screen MPD #A3, 185 ml - 0.1 M MES monohydrate pH 6.0, 10% (+/-)-2-Methyl-2,4-pentanediol
SDP
Supplier:  Hampton Research HR2930A3
Encompass
HR2-930-A3/Grid Screen MPD #A3, 185 ml - 0.1 M MES monohydrate pH 6.0, 10% (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-194-6