Learn More
Hampton Research HR2-928-A4/Grid Screen PEG/LiCl #A4, 185 ml - 0.1 M HEPES pH 7.0, 1.0 M Lithium chloride
SDP
Supplier:  Hampton Research HR2928A4
Encompass
HR2-928-A4/Grid Screen PEG/LiCl #A4, 185 ml - 0.1 M HEPES pH 7.0, 1.0 M Lithium chloride. Please note items from this supplier are non-returnable.
Catalog No. 50-192-3