Learn More
Hampton Research HR2-932-D5/Grid Screen Sodium Chloride #D5/Grid Screen Salt HT (H11), 185 ml - 0.1 M Tris pH 8.0, 4.0 M Sodium chloride
SDP
Supplier:  Hampton Research HR2932D5
Encompass
HR2-932-D5/Grid Screen Sodium Chloride #D5/Grid Screen Salt HT (H11), 185 ml - 0.1 M Tris pH 8.0, 4.0 M Sodium chloride. Please note items from this supplier are non-returnable.
Catalog No. 50-200-5