Learn More
Hampton Research HR2-932-C4/Grid Screen Sodium Chloride #C4/Grid Screen Salt HT (H04), 185 ml - 0.1 M HEPES pH 7.0, 3.0 M Sodium chloride
SDP
Supplier:  Hampton Research HR2932C4
Encompass
HR2-932-C4/Grid Screen Sodium Chloride #C4/Grid Screen Salt HT (H04), 185 ml - 0.1 M HEPES pH 7.0, 3.0 M Sodium chloride. Please note items from this supplier are non-returnable.
Catalog No. 50-199-8