Learn More
Hampton Research HR2-932-A6/Grid Screen Sodium Chloride #A6/Grid Screen Salt HT (G06), 185 ml - 0.1 M BICINE pH 9.0, 1.0 M Sodium chloride
SDP
Supplier:  Hampton Research HR2932A6
Encompass
HR2-932-A6/Grid Screen Sodium Chloride #A6/Grid Screen Salt HT (G06), 185 ml - 0.1 M BICINE pH 9.0, 1.0 M Sodium chloride. Please note items from this supplier are non-returnable.
Catalog No. 50-198-8