Learn More
Electron Microscopy Sciences Gilder Aperture Grid with Single 150 µm hole, Nickel
SDP
Supplier:  Electron Microscopy Sciences GA150NI
Encompass_Preferred
Single Slot aperture grids have a thickness of are 50 microns. Aperture diameter is 150 um.
Material: Nickel
Catalog No. 50-190-1285