Learn More
Electron Microscopy Sciences TEM Window Pure Silicon, (2) 100x1500 µm, 9nm
SDP
Catalog No. 501929467
Encompass_Preferred
Pure Silicon TEM Windows feature imaging windows with 5 to 15 nm thickness, reducing background contribution and interference for higher contrast imaging. The membrane can be vigorously plasma cleaned to remove organic contamination, unlike traditional carbon grids. The elemental silicon composition of TEM Windows remarkably increases stability at high beam currents and at high annealing temperatures. With a minimal background signal, elemental analyses of samples containing nitrogen and/or carbon is possible by EDX and EELS. Nanoporous with pores from 10-60 nm.
Dimensions: (2) 100x1500 µm
Thickness: 9nm
Catalog No. 50-192-9467 Supplier Electron Microscopy Sciences Supplier No. 7604275
Pure Silicon TEM Windows feature imaging windows with 5 to 15 nm thickness, reducing background contribution and interference for higher contrast imaging. The membrane can be vigorously plasma cleaned to remove organic contamination, unlike traditional carbon grids. The elemental silicon composition of TEM Windows remarkably increases stability at high beam currents and at high annealing temperatures. With a minimal background signal, elemental analyses of samples containing nitrogen and/or carbon is possible by EDX and EELS. Nanoporous with pores from 10-60 nm.
Dimensions: (2) 100x1500 µm
Thickness: 9nm