Learn More
Accuflow Systems Saint-Gobain Chemware Griffin Beaker, PTFE, 1 L, Low Form
Supplier: Accuflow Systems D1069041
The Chemware PTFE Griffin beaker is a 1,000 mL low-form laboratory vessel designed for handling highly corrosive reagents and applications that require chemically inert, non-contaminating containers. Constructed from PTFE (Teflon), the beaker is non-wettable and unbreakable, offering high chemical purity and resistance for demanding lab procedures.
- Capacity: 1,000 mL.
- Material: PTFE (Teflon).
- Beaker type: low-form Griffin beaker.
- Chemical resistance: inert to acids and bases, non-contaminating.
- Thermal: heat resistant (manufacturer lists use on hot plate up to 550°F / 288°C).
- Physical: non-wettable, lightweight, unbreakable.
- Handling: broad spout for easy pouring, specially molded bottom for rapid heat transfer.
- Maintenance: easy to clean, suitable for ultra-pure applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.