Learn More
Medchemexpress LLC Grifolic acid | 80557-12-6 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN39771MG
Encompass_Preferred
Grifolic acid is a phenolic compound initially isolated from the mushroom Albatrellus confluens. It functions as an agonist for the free fatty acid receptor (FFAR4/GPR120).
  • Acts as an agonist of the free fatty acid receptor (FFAR4/GPR120)
  • Reduces RAW264.7 macrophage cell viability
  • Exhibits antiproliferative activity against LPA-induced human DU-145 cells
Catalog No. 50-004-17948