Learn More
Medchemexpress LLC Griseofulvin | 126-07-8 | 98.9% | 352.77 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1758310MM/1ML
Encompass_Preferred
Griseofulvin is an orally active antifungal antibiotic with antitumor activity. It induces apoptosis and G2/M cell cycle arrest in cancer cells. Griseofulvin also exhibits cardiovascular modulatory activity, reducing angina pectoris, relieving hand artery spasm associated with onychomycosis, and peripheral vascular diseases such as shoulder-hand syndrome.
  • Orally active antifungal antibiotic
  • Exhibits antitumor activity
  • Induces apoptosis in cancer cells
  • Induces G2/M cell cycle arrest in cancer cells
  • Exhibits cardiovascular modulatory activity
  • Reduces angina pectoris
  • Relieves hand artery spasm
  • Relieves peripheral vascular diseases
Catalog No. 50-003-87397